Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WRB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553834
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553834 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553834 Supplier Invitrogen™ Supplier No. PA553834
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SFMSRVLQKD AEQESQMRAE IQDMKQELST VNMMDEFARY ARLERKINKM TDKLKTHVKA RTAQLAKIK Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

This gene encodes a basic nuclear protein of unknown function. The gene is widely expressed in adult and fetal tissues. Since the region proposed to contain the gene(s) for congenital heart disease (CHD) in Down syndrome (DS) patients has been restricted to 21q22.2-22.3, this gene, which maps to 21q22.3, has a potential role in the pathogenesis of Down syndrome congenital heart disease. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WRB
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GET1
Gene Accession No. O00258
Gene Alias 5530402J05Rik; C030018G21Rik; Chd5; Congenital heart disease 5 protein; GET1; tail-anchored protein insertion receptor WRB; tryptophan rich basic protein; tryptophan-rich basic protein; tryptophan-rich protein; Wrb
Gene Symbols GET1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human WRB. Recombinant protein control fragment (Product #RP-91126).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7485
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.