Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WRB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112787
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112787 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112787 Supplier Invitrogen™ Supplier No. PA5112787
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

WRB Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

This gene encodes a basic nuclear protein of unknown function. The gene is widely expressed in adult and fetal tissues. Since the region proposed to contain the gene(s) for congenital heart disease (CHD) in Down syndrome (DS) patients has been restricted to 21q22.2-22.3, this gene, which maps to 21q22.3, has a potential role in the pathogenesis of Down syndrome congenital heart disease. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WRB
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene GET1
Gene Accession No. O00258, Q8K0D7
Gene Alias 5530402J05Rik; C030018G21Rik; Chd5; Congenital heart disease 5 protein; GET1; tail-anchored protein insertion receptor WRB; tryptophan rich basic protein; tryptophan-rich basic protein; tryptophan-rich protein; Wrb
Gene Symbols GET1
Host Species Rabbit
Immunogen Recombinant Human Tail-anchored protein insertion receptor WRB protein (38-99AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 71446, 7485
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.