Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WRCH1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP133639

Catalog No. NBP133639


Only null left
Add to Cart

Description

Description

WRCH1 Polyclonal specifically detects WRCH1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

WRCH1
Polyclonal
Unconjugated
PBS (pH 7.0) and 10% Glycerol with 0.01% Thimerosal
ARHU, CDC42L1GTP-binding protein-like 1, DJ646B12.2, fJ646B12.2, FLJ10616,2310026M05Rik, G28K, GTP-binding protein like 1, GTP-binding protein SB128, hG28K, ras homolog gene family, member U, ras-like gene family member U, Rho GTPase-like protein ARHU, rho-related GTP-binding protein RhoU, Ryu GTPase, Wnt-1 responsive Cdc42 homolog 1, WRCH-1, WRCH1CDC42-like GTPase 1
Rabbit
28 kDa
100 μL
Cell Biology, Signal Transduction
58480
Human
IgG
Immunohistochemistry, Immunohistochemistry (Paraffin)
1.0mg/mL
Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Q7L0Q8
RHOU
Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human WRCH1. The exact sequence is proprietary.
Affinity Purified
RUO
Primary
Antibody reactive against peptide, detects immunogen. Further validation in progress.
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only