Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Wright' Stain (Powder/Certified Biological Stain), Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
Specifications
Specifications
| Bio. Stain Comm. Certification | Pass Test |
| Molecular Formula | C21H34O4 |
| MDL Number | MFCD00135237 |
| Identification | Pass Test |
| InChI Key | DJKDIKIDYDXHDD-IGUVKOCZSA-N |
| SMILES | CCCCC[C@H](O)\C=C\[C@H]1[C@@H]2CO[C@@H](C2)[C@@H]1C\C=C/CCCC(O)=O |
| IUPAC Name | (5Z)-7-[(1S,4R,5S,6R)-5-[(1E,3S)-3-hydroxyoct-1-en-1-yl]-2-oxabicyclo[2.2.1]heptan-6-yl]hept-5-enoic acid |
| Molecular Weight (g/mol) | 350.50 |
| PubChem CID | 5311386 |
| Grade | Certified Biological Stain |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction