Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ Wrinkle Textured Latex Coated Cotton Gloves

Catalog No. 19014962 Shop All Showa Best Glove Products
Change view
Click to view available options
Back Material:
Cotton
Elasticated
Size:
Small
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Back Material Size
19-014-962 Cotton Small
19-806-658 Elasticated Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-014-962 Supplier SHOWA™ Supplier No. L66NFW08
Add to Cart
Add to Cart
This item is not returnable. View return policy

Gloves provide heavy duty protection with a natural latex coating for an enhanced grip in wet or dry conditions. The soft liner provides comfort and flexibility, while a reinforced safety cuff offers additional protection.

  • Heavy duty protection
  • Fully coated with natural latex
  • Texturized finish for enhanced grip
  • Cotton flannel liner increases comfort
  • Optional elasticated back for a secure fit
  • Reinforced safety cuff provides additional protection
  • In compliance 21 CFR 177.2600

Specifications

Back Material Cotton
Certifications/Compliance FDA 21 CFR 177.2600
Coated Area Palm, Fingers
Cuff Material Reinforced Cotton
Coating Material Latex
Color Yellow
Exterior Finish Fully Textured
Hand Specific
Material Cotton
Product Type Dip-coated Gloves
Size Small
For Use With (Application) Landscaping, Wood Cutting, Light Chemical Handling, and General Maintenance
Glove Cuff Color Natural
Glove Cuff Style Safety
Liner Material Flannel
Series L66NFW
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.