Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WRN Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100472G
Description
WRN Polyclonal antibody specifically detects WRN in Human, Mouse samples. It is validated for Western BlotSpecifications
WRN | |
Polyclonal | |
Western Blot | |
DKFZp686C2056, DNA helicase, RecQ-like type 3, EC 3.1, EC 3.6.1, EC 3.6.4.12, Exonuclease WRN, RecQ protein-like 2, RecQ3, RECQ3RECQL3, RECQL2Werner syndrome ATP-dependent helicase, Werner syndrome, Werner syndrome, RecQ helicase-like | |
The immunogen recognized by this antibody maps to a region between residues 1400 and the C-terminus (residue 1432) of human Werner Syndrome Helicase using the numbering given in SwissProt entry Q14191 (GeneID 7486). | |
0.1 mL | |
Cancer, DNA Repair, Genes Sensitive to DNA Damaging Agents, Tumor Suppressors | |
7486 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction