Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WRP-50 All-Hazards Alert (NOAA) Radio
SDPSCI_ED

Catalog No. s94726 Shop All Life Secure Products
Change view
Click to view available options
:
All-Hazards Alert (NOAA) Radio; 120V with 9V battery backup
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S94726 All-Hazards Alert (NOAA) Radio; 120V with 9V battery backup
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S94726 Supplier Life Secure Supplier No. 70900
Only null left
Add to Cart
Add to Cart

Turns on automatically to supply instant alerts to dangerous situations.

  • Broadcasts warnings plus information from the government on disaster precautions and procedures
  • Specific Area Message Encoding (S.A.M.E.) eliminates false alerts
  • Recommended by the U.S. government for all public/private facilities
  • Provides 24-hour weather updates at the touch of a button
  • Mounts on desk or wall
  • 120V with 9V battery backup
  • Dimensions: 11.5L x 6.5W x 4 in.H (38.1 x 16.5 x 10.2cm)

Specifications

Height (English) 4 in.
Height (Metric) 10.2 cm
Voltage 120 V
Width (English) 6.5 in.
Width (Metric) 16.5 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.