Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WSB1 Antibody (K5P2H10*E10), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250365F
Description
WSB1 Monoclonal specifically detects WSB1 in Human samples. It is validated for ELISA, Immunohistochemistry.Specifications
WSB1 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
WSB1 | |
Ovalbumin-conjugated synthetic peptide RRVMPTQEVQE | |
0.1 mL | |
Signal Transduction | |
26118 | |
Store at 4C in the dark. | |
IgG1 κ |
ELISA, Immunohistochemistry | |
K5P2H10*E10 | |
ELISA, Immunohistochemistry | |
DKFZp564B0482, SOCS box-containing WD protein SWiP-1, SWIP1DKFZp564A122, WD repeat and SOCS box containing 1, WD repeat and SOCS box-containing 1, WD repeat and SOCS box-containing protein 1, WSB-1 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction