Learn More
Invitrogen™ WSB1 Monoclonal Antibody (3E10)

Description
Peptide Sequence: QGHRTVKLVP WSQCLQNFLL HGTKNVTNSS SLRLPRQNSD GGQKNKPREH IIDCGDIVWS LAFGSSVPEK QSRCVNIEWH RFRFG.
Specifications
Specifications
| Antigen | WSB1 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 3E10 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | WSB1 |
| Gene Accession No. | Q9Y6I7 |
| Gene Alias | 1110056B13Rik; 2700038M07Rik; SOCS box-containing WD protein SWiP-1; SWIP1; WD repeat and SOCS box containing 1; WD repeat and SOCS box-containing 1; WD repeat and SOCS box-containing protein 1; WD repeat and SOCS box-containing protein 1-like protein; WD-40-repeat-containing protein with a SOCS box 1; WSB1; WSB-1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.