Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WSB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551884
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551884 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551884 Supplier Invitrogen™ Supplier No. PA551884
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: APFDKKCGRE NWTVAFAPDG SYFAWSQGHR TVKLVPWSQC LQNFLLHGTK NVTNSSSLRL PRQNSDGGQK NKPREHIIDC GDIVWSLAFG SSVPEKQSRC VNIEWHRFRF GQDQLLLATG LNNGRIKIWD V Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 93%.

WSB1 (WD repeat and SOCS box-containing 1), also known as SWIP1, is a 421 amino acid protein that contains one SOCS box domain and six WD repeats. Expressed as multiple alternatively spliced isoforms, WSB1 is thought to function as part of an SCF-like ECS (Elongin-Cullin-SOCS-box protein) E3 ubiquitin ligase complex that mediates the ubiquitination and proteasomal degradation of target proteins, such as DIO2. Overexpression of WSB1 is implicated in pancreatic cancer progression, suggesting a role for WSB1 in carcinogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WSB1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WSB1
Gene Accession No. Q9Y6I7
Gene Alias 1110056B13Rik; 2700038M07Rik; SOCS box-containing WD protein SWiP-1; SWIP1; WD repeat and SOCS box containing 1; WD repeat and SOCS box-containing 1; WD repeat and SOCS box-containing protein 1; WD repeat and SOCS box-containing protein 1-like protein; WD-40-repeat-containing protein with a SOCS box 1; WSB1; WSB-1
Gene Symbols WSB1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human WSB1. Recombinant protein control fragment (Product #RP-102067).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26118
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.