Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ WSB1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA575162
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
WSB1 (WD repeat and SOCS box-containing 1), also known as SWIP1, is a 421 amino acid protein that contains one SOCS box domain and six WD repeats. Expressed as multiple alternatively spliced isoforms, WSB1 is thought to function as part of an SCF-like ECS (Elongin-Cullin-SOCS-box protein) E3 ubiquitin ligase complex that mediates the ubiquitination and proteasomal degradation of target proteins, such as DIO2. Overexpression of WSB1 is implicated in pancreatic cancer progression, suggesting a role for WSB1 in carcinogenesis.
Specifications
WSB1 | |
Polyclonal | |
Unconjugated | |
WSB1 | |
1110056B13Rik; 2700038M07Rik; SOCS box-containing WD protein SWiP-1; SWIP1; WD repeat and SOCS box containing 1; WD repeat and SOCS box-containing 1; WD repeat and SOCS box-containing protein 1; WD repeat and SOCS box-containing protein 1-like protein; WD-40-repeat-containing protein with a SOCS box 1; WSB1; WSB-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26118, 303336, 78889 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.05% sodium azide; pH 7.3 | |
O54927, Q9Y6I7 | |
WSB1 | |
Recombinant full length Human WSB1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction