Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ WSCD2 Monoclonal Antibody (9-F3)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA532883
Description
WSCD2 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
The function remains known.
Specifications
WSCD2 | |
Monoclonal | |
2 mg/mL | |
PBS with 0.2% BSA, 50% Glycerol and 0.05% sodium azide; pH 7.4 | |
Q2TBF2 | |
WSCD2 | |
Recombinant protein with Human WSCD2 50-353. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgM |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
9-F3 | |
Unconjugated | |
WSCD2 | |
4933413A10Rik; C530024P05Rik; Gm450; KIAA0789; MGC117165; si:ch211-240b21.1; WSC domain containing 2; WSC domain containing 2, pseudogene 1; WSC domain-containing protein 2; wscd2; Wscd2-ps1 | |
Mouse | |
Protein L | |
RUO | |
9671 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction