Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WSCD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553046
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553046 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553046 Supplier Invitrogen™ Supplier No. PA553046
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GAELSFLGDM HLGRGFRDTG EASSIARRYG PWFKGKDGNE RAKLGDYGGA WSRALKGRVV REKEEERAKY IGCYLDDTQS RALRGVSFFD YKKMTIFRCQ DNCAERGYLY Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.

The function remains known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WSCD2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WSCD2
Gene Accession No. Q2TBF2
Gene Alias 4933413A10Rik; C530024P05Rik; Gm450; KIAA0789; MGC117165; si:ch211-240b21.1; WSC domain containing 2; WSC domain containing 2, pseudogene 1; WSC domain-containing protein 2; wscd2; Wscd2-ps1
Gene Symbols WSCD2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human WSCD2. Recombinant protein control fragment (Product #RP-89984).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9671
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.