Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WSTF Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP303399100UL
Description
WSTF Polyclonal specifically detects WSTF in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated.Specifications
WSTF | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence, KnockDown | |
Unconjugated | |
bromodomain adjacent to zinc finger domain, 1B, williams-Beuren syndrome chromosomal region 10 protein | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human WSTF (NP_115784.1). | |
100 μL | |
Transcription Factors and Regulators | |
9031 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction