Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WT1 Antibody (6F-H2) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 5 publications

$203.95 - $556.85

Specifications

Antigen WT1
Clone 6F-H2
Concentration 1.0 mg/mL
Dilution Western Blot 2 ug/ml, Simple Western 1:50, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 25576053), Immunoprecipitation 2-10 ug/ml lysate, Immunohistochemistry-Paraffin 1:400, Immunohistochemistry-Frozen 1: 400
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB11060011T
SDP
View Documents
Novus Biologicals
NB11060011SS
0.025 mL
Each for $203.95
Only null left
Add to Cart
 
NB11060011
SDP
View Documents
Novus Biologicals
NB11060011
0.1 mL
Each for $556.85
Only null left
Add to Cart
 
Description

Description

WT1 Monoclonal specifically detects WT1 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

WT1
1.0 mg/mL
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Unconjugated
Mouse
Angiogenesis, Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Tumor Suppressors
P19544
7490
Human recombinant Wilms Tumor 1 protein (WT1) (residues 1-181). [Swiss-Prot: P19544]
Primary
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
6F-H2
Western Blot 2 ug/ml, Simple Western 1:50, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 25576053), Immunoprecipitation 2-10 ug/ml lysate, Immunohistochemistry-Paraffin 1:400, Immunohistochemistry-Frozen 1: 400
Monoclonal
Purified
RUO
Human, Mouse
AWT1, NPHS4GUD, WAGR, Wilms tumor 1, Wilms tumor protein, WIT-2, WT33
WT1
IgG1 κ
Protein G purified
55 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.