Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WT1 Antibody (rWT1/857), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$257.16 - $554.51

Specifications

Antigen WT1
Clone rWT1/857
Concentration 0.2 mg/mL
Dilution Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB25318120
SDP
View Documents
Novus Biologicals
NBP25318120UG
20 μg
Each for $257.16
Only null left
Add to Cart
 
NB253181100
SDP
View Documents
Novus Biologicals
NBP253181100UG
100 μg
Each for $554.51
Only null left
Add to Cart
 
NB253181200
SDP
View Documents
Novus Biologicals
NBP253181200UG
200 μg N/A N/A N/A
Description

Description

WT1 Monoclonal specifically detects WT1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
Specifications

Specifications

WT1
0.2 mg/mL
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Angiogenesis, Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Tumor Suppressors
1mM PBS with 0.05% BSA with 0.05% Sodium Azide
7490
Recombinant full-length human WT1 protein
Primary
Store at 4C.
rWT1/857
Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml
Monoclonal
Purified
RUO
Human
AWT1, NPHS4GUD, WAGR, Wilms tumor 1, Wilms tumor protein, WIT-2, WT33
WT1
IgG1 κ
Protein A or G purified
51 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.