Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WT1 Antibody (SPM361), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311553AF594
Description
WT1 Monoclonal antibody specifically detects WT1 in Human, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
WT1 | |
Monoclonal | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
SPM361 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
AWT1, NPHS4GUD, WAGR, Wilms tumor 1, Wilms tumor protein, WIT-2, WT33 | |
Recombinant protein corresponding to residues 1-181 of human WT1. (Uniprot: P19544) | |
0.1 mL | |
Angiogenesis, Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Tumor Suppressors | |
7490 | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction