Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WT1 Antibody (WT1/1434R), Novus Biologicals™

Rabbit Monoclonal Antibody
$267.77 - $566.53
Specifications
| Antigen | WT1 |
|---|---|
| Clone | WT1/1434R |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml |
| Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB25318220
![]() |
Novus Biologicals
NBP25318220UG |
20 μg |
Each for $267.77
|
|
|||||
NB253182100
![]() |
Novus Biologicals
NBP253182100UG |
100 μg |
Each for $566.53
|
|
|||||
NB253182200
![]() |
Novus Biologicals
NBP253182200UG |
200 μg | N/A | N/A | N/A | ||||
Description
WT1 Monoclonal specifically detects WT1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| WT1 | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Tumor Suppressors | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| 7490 | |
| Recombinant human full-length WT1 protein | |
| Primary | |
| Store at 4C. |
| WT1/1434R | |
| Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| AWT1, NPHS4GUD, WAGR, Wilms tumor 1, Wilms tumor protein, WIT-2, WT33 | |
| WT1 | |
| IgG κ | |
| Protein A or G purified | |
| 51 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title