Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WT1 Antibody (WT1/3477R) - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308966
Description
WT1 Monoclonal specifically detects WT1 in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
WT1 | |
Monoclonal | |
1 mg/mL | |
Immunohistochemistry-Paraffin : 1-2 ug/mL | |
AWT1, NPHS4GUD, WAGR, Wilms tumor 1, Wilms tumor protein, WIT-2, WT33 | |
Synthetic peptide corresponding to WT1 residues (within aa1-100) of WT1 | |
100 μg | |
Angiogenesis, Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Tumor Suppressors | |
7490 | |
Store at -20 to -80C. Avoid freeze-thaw cycles. | |
IgG κ |
Immunohistochemistry (Paraffin) | |
WT1/3477R | |
Unconjugated | |
10mM PBS | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction