Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WT1 Antibody (WT1/857) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$646.00
Specifications
Antigen | WT1 |
---|---|
Clone | WT1/857 |
Concentration | 1.0 mg/mL |
Dilution | Flow Cytometry : 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin : 0.5 - 1.0 ug/ml, Immunofluorescence : 0.5 - 1.0 ug/ml, CyTOF-ready |
Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP247931A
![]() |
Novus Biologicals
NBP2479310.1MG |
0.1 mg |
Each for $646.00
|
|
|||||
NBP247931B
![]() |
Novus Biologicals
NBP2479310.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
WT1 Monoclonal specifically detects WT1 in Human, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
WT1 | |
1.0 mg/mL | |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF | |
Unconjugated | |
Mouse | |
Angiogenesis, Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Tumor Suppressors | |
PBS with No Preservative | |
7490 | |
Recombinant human WT1 protein | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
WT1/857 | |
Flow Cytometry : 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin : 0.5 - 1.0 ug/ml, Immunofluorescence : 0.5 - 1.0 ug/ml, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
AWT1, NPHS4GUD, WAGR, Wilms tumor 1, Wilms tumor protein, WIT-2, WT33 | |
WT1 | |
IgG1 κ | |
Protein A or G purified | |
Recognizes a 47-55kDa-tumor suppressor protein, identified as Wilm's Tumor (WT1) protein. The antibody reacts with all isoforms of the full-length WT1 and also identifies WT1 lacking exon 2-encoded amino acids, frequently found in subsets of sporadic Wilm s tumors.WT1, a sporadic and familial pediatric kidney tumor, is genetically heterogeneous. Wilm s tumor is associated with mutations of WT1, a zinc-finger transcription factor that is essential for the development of the metanephric kidney and the urogenital system. The WT1 gene is normally expressed in fetal kidney and mesothelium, and its expression has been suggested as a marker for Wilm s tumor and mesothelioma. WT1 protein has been identified in proliferative mesothelial cells, malignant mesothelioma, ovarian carcinoma, gonadoblastoma, nephroblastoma, and desmoplastic small round cell tumor. Lung adenocarcinomas rarely stain positive with this antibody. WT1 protein expression in mesothelial cells has become a reliable marker for the diagnosis of mesotheliomas. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title