Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WT1-AS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575590
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575590 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575590 Supplier Thermo Scientific Supplier No. PA575590
Add to Cart
Add to Cart

WT1-AS Polyclonal Antibody

WT1-AS Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen WT1-AS
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene WT1-AS
Gene Accession No. Q06250
Gene Alias 6030498F17; AI314831; Wilms tumor 1 homolog, opposite strand; WIT1; WIT-1; WT1 antisense RNA; WT1AS; Wt1-as; WT1-AS1; Wt1os
Gene Symbols WT1-AS, Wt1os
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues in Human WT1-AS.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 329480, 51352
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.