Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WT1 Mouse anti-Human, Mouse, Rat, Clone: WT1/857, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB14650
Description
Mouse monoclonal antibody raised against full length recombinant human WT1.
- High specificity
- High reactivity
Specifications
WT1 | |
Monoclonal | |
Unconjugated | |
Flow Cytometry (0.5-1 ug/million cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
P19544 | |
WT1 | |
Recombinant protein corresponding to full length human WT1. | |
100 μg | |
Primary | |
The antibody reacts with all isoforms of the full length WT1 and also identifies WT1 lacking exon 2 encoded amino acids. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 κ |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
WT1/857 | |
Mouse monoclonal antibody raised against full length recombinant human WT1. | |
In PBS. | |
GUD/WAGR/WIT-2/WT33 | |
Mouse | |
Protein G purification | |
RUO | |
7490 | |
Human, Mouse, Rat | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction