Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WT1-AS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103316
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103316 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103316 Supplier Invitrogen™ Supplier No. PA5103316
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total WIT1.

Wilms' tumor is a neoplasm of the kidneys that usually occurs in children and is characterized by the presence of abortive tubules and glomeruli surrounded by a spindled cell stroma. The 11p13 Wilms' tumor locus consists of two coordinately regulated transcripts, WT1 and WIT-1, which are mutated in Wilms' tumors. Wilms' tumor upstream neighbor 1 (WIT-1) is encoded by an intron-less gene upstream of the Wilms' tumor 1 (WT1) gene, which is is important for nephrogenesis and gonadal growth. The WT1 gene is bi-directionally transcribed from the same promoter region as WIT-1, which may function as an antisense regulator of WT1. WIT-1 and WT1 have the same temporal and cell-restricted expression pattern, although the expression of WIT-1 is less abundant. Methylation of the WIT-1 gene is implicated in hematologic malignancy of chemoresistant acute myeloid leukemia. Single nucleotide polymorphisms (SNPs) in the WIT-1 gene are significantly associated with focal segmental glomerulosclerosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WT1-AS
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene WT1-AS
Gene Accession No. Q06250
Gene Alias 6030498F17; AI314831; Wilms tumor 1 homolog, opposite strand; WIT1; WIT-1; WT1 antisense RNA; WT1AS; Wt1-as; WT1-AS1; Wt1os
Gene Symbols WT1-AS, Wt1os
Host Species Rabbit
Immunogen A synthesized peptide derived from human WT1-AS(Accession Q06250), corresponding to amino acid residues V12-E62.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 329480, 51352
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.