Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WT1 Rabbit anti-Human, Clone: WT1/3477R, Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

$316.50

Specifications

Antigen WT1
Clone WT1/3477R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB121916
SDP
View Documents
Novus Biologicals
NBP30722120UG
20 μg
Each of 1 for $316.50
Only null left
Add to Cart
 
Description

Description

WT1 Monoclonal specifically detects WT1 in Human samples. It is validated for Immunohistochemistry-Paraffin.
Specifications

Specifications

WT1
0.2 mg/ml
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Angiogenesis, Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Tumor Suppressors
10 mM PBS with 0.05% BSA
7490
IgG κ
Protein A or G purified
WT1/3477R
Immunohistochemistry-Paraffin 1-2 ug/ml
Monoclonal
Purified
RUO
Human
AWT1, NPHS4GUD, WAGR, Wilms tumor 1, Wilms tumor protein, WIT-2, WT33
Synthetic peptide corresponding to WT1 residues (within aa1-100) of WT1
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.