Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WTAP Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299848JF646
Description
WTAP Polyclonal antibody specifically detects WTAP in Human samples. It is validated for Western Blot, ELISASpecifications
WTAP | |
Polyclonal | |
Western Blot, ELISA | |
Female-lethal(2)D homolog, hFL(2)D, KIAA0105DKFZp686F20131, MGC3925, PNAS-132, pre-mRNA-splicing regulator WTAP, putative pre-mRNA splicing regulator female-lethal(2D), Wilms tumor 1 associated protein, Wilms tumor 1-associating protein, Wilms' tumour 1-associating protein, WT1-associated protein | |
Produced in rabbits immunized with purified, recombinant Human WTAP (Uniprot#: Q15007-2; Met 1-Arg 151) | |
0.1 mL | |
Cancer, Tumor Suppressors | |
9589 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction