Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WTAP Antibody (OTI12G7), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274870JF646
Description
WTAP Monoclonal specifically detects WTAP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
WTAP | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
WTAP | |
Recombinant protein fragment corresponding to amino acids 2-106 of human WTAP (NP_004897) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI12G7 | |
Western Blot, Immunohistochemistry | |
Female-lethal(2)D homolog, hFL(2)D, KIAA0105DKFZp686F20131, MGC3925, PNAS-132, pre-mRNA-splicing regulator WTAP, putative pre-mRNA splicing regulator female-lethal(2D), Wilms tumor 1 associated protein, Wilms tumor 1-associating protein, Wilms′ tumour 1-associating protein, WT1-associated protein | |
Mouse | |
Affinity Purified | |
Cancer, Tumor Suppressors | |
9589.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction