Learn More
WTAP Mouse anti-Human, Clone: OTI6H3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF808316

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The Wilms tumor suppressor gene WT1 appears to play a role in both transcriptional and posttranscriptional regulation of certain cellular genes. This gene encodes a WT1-associating protein, which is a ubiquitously expressed nuclear protein. Like WT1 protein, this protein is localized throughout the nucleoplasm as well as in speckles and partially colocalizes with splicing factors. Alternative splicing of this gene results in three transcript variants, two of which encode the same isoform.Specifications
WTAP | |
Monoclonal | |
Unconjugated | |
WTAP | |
Mum2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
9589 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI6H3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q15007 | |
WTAP | |
Human recombit protein fragment corresponding to amino acids 2-106 of human WTAP produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.