Learn More
Invitrogen™ WTAP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589988
Description
Immunogen sequence: DEEVEGMQST ILVLQQQLKE TRQQLAQYQQ QQSQASAPST SRTTASEPVE QSEATSKDCS RLTNGPSNGS SSRQRTSGSG FHREGNTTED DFPSSPGNGN KSSNSSEERT GRGGSGYVNQ LSAGYESVDS PTGSENSLTH QSND Positive Samples: Rat spleen; Cellular Location: Nucleus, Nucleus speckle, nucleoplasm.
Wilms' tumor (WT) is an embryonal malignancy of the kidney that affects 1 in 10,000 infants and is observed in both sporadic and inherited forms. The Wilms' tumor protein (WT1) binds the DNA sequence GCGGGGGCG, a recognition element common to the early growth response (Egr) family of Zn2+ finger transcriptional activators, and functions as a transcriptional repressor. WTAP (Wilms tumor 1-associating protein) is a ubiquitously expressed nuclear protein that interacts with WT1 and may be involved in regulating mRNA splicing. WTAP is found in nuclear speckles, where it regulates the G2/M cell cycle transition by binding to the 3' UTR of cyclin A2, thus enhancing its stability. Additionally, WTAP inhibits expression of WT1 target genes and is able to impair the ability of WT1 to bind DNA. Two isoforms of WTAP exist due to alternative splicing events.
Specifications
WTAP | |
Polyclonal | |
Unconjugated | |
WTAP | |
2810408K05Rik; 9430038B09Rik; Female-lethal(2)D homolog; hFL(2)D; KIAA0105; Mum2; PNAS-132; pre-mRNA-splicing regulator WTAP; putative pre-mRNA splicing regulator female-lethal(2D); RGD1563824; Wilms tumor 1 associated protein; wilms tumor 1-associating protein; Wilms' tumour 1-associating protein; Wilms tumour 1-associating protein; WT1-associated protein; WTAP | |
Rabbit | |
Affinity Chromatography | |
RUO | |
499020, 60532, 9589 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q15007, Q9ER69 | |
WTAP | |
Recombinant fusion protein containing a sequence corresponding to amino acids 212-355 of human WTAP (NP_0048972). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.