Learn More
WTAP Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301435AT
Description
The recommended shelf life for this product is 1 year from date of receipt.
The Wilms tumor suppressor gene WT1 appears to play a role in both transcriptional and posttranscriptional regulation of certain cellular genes. This gene encodes a WT1-associating protein, which is a ubiquitously expressed nuclear protein. Like WT1 protein, this protein is localized throughout the nucleoplasm as well as in speckles and partially colocalizes with splicing factors. Alternative splicing of this gene results in three transcript variants, two of which encode the same isoform.Specifications
| WTAP | |
| Polyclonal | |
| Unconjugated | |
| WTAP | |
| PNAS-132, WT1-associated protein, Wilms′ tumour 1-associating protein, female-lethal(2)D homolog, hFL(2)D, pre-mRNA-splicing regulator WTAP, putative pre-mRNA splicing regulator female-lethal(2D), wilms tumor 1-associating protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9589 | |
| 4° C | |
| Liquid |
| Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q15007 | |
| WTAP | |
| Between 250 and 300 | |
| 10 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.