Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WWC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571284
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571284 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571284 Supplier Invitrogen™ Supplier No. PA571284
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 92%; Dog: 100%; Guinea Pig: 92%; Horse: 93%; Human: 100%; Mouse: 92%; Pig: 100%; Rat: 92%; Yeast: 83%.

This gene encodes a member of the WW-and-C2-domain-containing family of proteins. Members of this family have two N-terminal WW domains that mediate binding to target proteins harboring L/PPxY motifs, an internal C2 domain for membrane association, and C-terminal alpha protein kinase C binding sites and class III PDZ domain-interaction motifs. Proteins of this family are able to form homo- and heterodimers and to modulate hippo pathway signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WWC2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene WWC2
Gene Accession No. Q6AWC2
Gene Alias 5330431C14Rik; AU022508; BH-3-only member B; BH3-only member B protein; BOMB; C85457; D8Ertd594e; LOW QUALITY PROTEIN: protein WWC2; MNCb-4173; Protein WWC2; RGD1559427; WW and C2 domain containing 2; WW domain-containing protein 2; WW, C2 and coiled-coil domain containing 2; WWC2
Gene Symbols WWC2
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of human WWC2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80014
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.