Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WWC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP315585100UL
Description
WWC2 Polyclonal antibody specifically detects WWC2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| WWC2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| C2 and coiled-coil domain containing 2, FLJ34082, WW and C2 domain containing 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 160-430 of human WWC2 (NP_079225.5). DPDILKAEISTTRLRVKKLKRELSQMKQELLYKEQGFETLQQIDKKMSGGQSGYELSEAKAILTELKSIRKAISSGEKEKQDLMQSLAKLQERFHLDQNIGRSEPDLRCSPVNSHLCLSRQTLDAGSQTSISGDIGVRSRSNLAEKVRLSLQYEEAKRSMANLKIELSKLDSEAWPGALDIEKEKLMLINEKEELLKELQFVTPQKRTQDELERLEAERQRLEEELLSVRGTPSRALAERLRLEERRKELLQKLEETTKLTTYLHSQLKSL | |
| 100 μg | |
| Cell Biology | |
| 80014 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction