Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WWC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP31558520UL
This item is not returnable.
View return policy
Description
WWC2 Polyclonal antibody specifically detects WWC2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
WWC2 | |
Polyclonal | |
Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
C2 and coiled-coil domain containing 2, FLJ34082, WW and C2 domain containing 2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 160-430 of human WWC2 (NP_079225.5). DPDILKAEISTTRLRVKKLKRELSQMKQELLYKEQGFETLQQIDKKMSGGQSGYELSEAKAILTELKSIRKAISSGEKEKQDLMQSLAKLQERFHLDQNIGRSEPDLRCSPVNSHLCLSRQTLDAGSQTSISGDIGVRSRSNLAEKVRLSLQYEEAKRSMANLKIELSKLDSEAWPGALDIEKEKLMLINEKEELLKELQFVTPQKRTQDELERLEAERQRLEEELLSVRGTPSRALAERLRLEERRKELLQKLEETTKLTTYLHSQLKSL | |
20 μg | |
Cell Biology | |
80014 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction