Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WWC3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558941
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558941 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558941 Supplier Invitrogen™ Supplier No. PA558941
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KRLERRARRI SACLSDYSLA SDSGVFEPLT KRNEDAEEPA YGDTASNGDP QIHVGLLRDS GSECLLVHVL QLKNPAGLAV KEDCKVHIRV YLPPLDSGT Highest antigen sequence identity to the following orthologs: Mouse - 31%, Rat - 82%.

This gene encodes a member of the WWC family of proteins, which also includes the WWC1 (KIBRA) gene product and the WWC2 gene product. The protein encoded by this gene includes a C2 domain, which is known to mediate homodimerization in the related WWC1 gene product.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WWC3
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WWC3
Gene Accession No. Q9ULE0
Gene Alias BM042; KIAA1280; Protein WWC3; RGD1560666; WWC family member 3; Wwc3
Gene Symbols WWC3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human WWC3. Recombinant protein control fragment (Product #RP-98317).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55841
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.