Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WWOX Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA517237
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA517237 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA517237 Supplier Invitrogen™ Supplier No. PA517237
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

It is not recommended to aliquot this antibody.

WW domain-containing proteins are found in all eukaryotes and play an important role in the regulation of a wide variety of cellular functions such as protein degradation, transcription, and RNA splicing. This gene encodes a protein which contains 2 WW domains and a short-chain dehydrogenase/reductase domain (SRD). The highest normal expression of this gene is detected in hormonally regulated tissues such as testis, ovary, and prostate. This expression pattern and the presence of an SRD domain suggest a role for this gene in steroid metabolism. The encoded protein is more than 90% identical to the mouse protein, which is an essential mediator of tumor necrosis factor-alpha-induced apoptosis, suggesting a similar, important role in apoptosis for the human protein. In addition, there is evidence that this gene behaves as a suppressor of tumor growth. Alternative splicing of this gene generates transcript variants that encode different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WWOX
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 0.01M HEPES with 100 μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene Wwox
Gene Accession No. Q91WL8, Q9NZC7
Gene Alias 5330426P09Rik; 9030416C10Rik; D16S432E; EIEE28; FOR; FRA16D; Fragile site FRA16D oxidoreductase; HHCMA56; PRO0128; SCAR12; SDR41C1; Short chain dehydrogenase/reductase family 41C member 1; short chain dehydrogenase/reductase family 41C, member 1; WOX1; WW domain containing oxidoreductase; WW domain-containing oxidoreductase; WW domain-containing protein WWOX; WW-domain oxidoreductase; Wwox
Gene Symbols Wwox
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues surrounding Thr103 of WWOX.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 292041, 51741, 80707
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.