Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WWP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554715
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554715 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554715 Supplier Invitrogen™ Supplier No. PA554715
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TIDLKQALLI HNRKLERVKE QLKLSLENKN GIAQTGELTV VLDGLVIEQE NITNCSSSPT IEIQENGDAL HENGEPSART TARLAVEGTN GIDNHV Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 82%.

WW domain-containing proteins are found in all eukaryotes and play an important role in the regulation of a wide variety of cellular functions such as protein degradation, transcription, and RNA splicing. This gene encodes a protein which contains 4 tandem WW domains and a HECT (homologous to the E6-associated protein carboxyl terminus) domain. The encoded protein belongs to a family of NEDD4-like proteins, which are E3 ubiquitin-ligase molecules and regulate key trafficking decisions, including targeting of proteins to proteosomes or lysosomes. Alternative splicing of this gene generates at least 6 transcript variants; however, the full length nature of these transcripts has not been defined. [provided by RefSeq].
TRUSTED_SUSTAINABILITY

Specifications

Antigen WWP1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Wwp1
Gene Accession No. Q9H0M0
Gene Alias 8030445B08Rik; AIP5; atrophin-1 interacting protein 5; atrophin-1-interacting protein 5; DKFZp434D2111; HECT-type E3 ubiquitin transferase WWP1; hSDRP1; NEDD4-like E3 ubiquitin-protein ligase WWP1; Nedd-4-like ubiquitin-protein ligase; SDRP1; TGIF-interacting ubiquitin ligase 1; Tiul1; Unknown (protein for MGC:129005); WW domain containing E3 ubiquitin protein ligase 1; WW domain-containing protein 1; Wwp1
Gene Symbols Wwp1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human WWP1. Recombinant protein control fragment (Product #RP-93779).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11059
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.