Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ WWP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5106453
Description
Antibody detects endogenous levels of total WWP1.
WW domain-containing proteins are found in all eukaryotes and play an important role in the regulation of a wide variety of cellular functions such as protein degradation, transcription, and RNA splicing. This gene encodes a protein which contains 4 tandem WW domains and a HECT (homologous to the E6-associated protein carboxyl terminus) domain. The encoded protein belongs to a family of NEDD4-like proteins, which are E3 ubiquitin-ligase molecules and regulate key trafficking decisions, including targeting of proteins to proteosomes or lysosomes. Alternative splicing of this gene generates at least 6 transcript variants; however, the full length nature of these transcripts has not been defined. [provided by RefSeq].
Specifications
WWP1 | |
Polyclonal | |
Unconjugated | |
Wwp1 | |
8030445B08Rik; AIP5; atrophin-1 interacting protein 5; atrophin-1-interacting protein 5; DKFZp434D2111; HECT-type E3 ubiquitin transferase WWP1; hSDRP1; NEDD4-like E3 ubiquitin-protein ligase WWP1; Nedd-4-like ubiquitin-protein ligase; SDRP1; TGIF-interacting ubiquitin ligase 1; Tiul1; Unknown (protein for MGC:129005); WW domain containing E3 ubiquitin protein ligase 1; WW domain-containing protein 1; Wwp1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
107568, 11059, 297930 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8BZZ3, Q9H0M0 | |
Wwp1 | |
A synthesized peptide derived from human WWP1(Accession Q9H0M0), corresponding to amino acid residues R508-C558. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction