Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WWP2 Antibody (02), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306109PCP
Description
WWP2 Monoclonal antibody specifically detects WWP2 in Human samples. It is validated for ELISASpecifications
WWP2 | |
Monoclonal | |
PerCP | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
02 | |
ELISA | |
AIP2Nedd-4-like ubiquitin-protein ligase, atrophin-1 interacting protein 2, Atrophin-1-interacting protein 2, EC 6.3.2, EC 6.3.2.-, NEDD4-like E3 ubiquitin-protein ligase WWP2, WW domain containing E3 ubiquitin protein ligase 2, WW domain-containing protein 2, WWp2-like | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human WWP2 (Uniprot#: O00308; Met1-Glu870). | |
0.1 mL | |
Cell Biology | |
11060 | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction