Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WWP2 Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP149942G
Description
WWP2 Polyclonal antibody specifically detects WWP2 in Human, Mouse samples. It is validated for Western BlotSpecifications
WWP2 | |
Polyclonal | |
Western Blot | |
AIP2Nedd-4-like ubiquitin-protein ligase, atrophin-1 interacting protein 2, Atrophin-1-interacting protein 2, EC 6.3.2, EC 6.3.2.-, NEDD4-like E3 ubiquitin-protein ligase WWP2, WW domain containing E3 ubiquitin protein ligase 2, WW domain-containing protein 2, WWp2-like | |
The immunogen for this product maps to a region between residue 200 and 250 of human WW Domain Containing E3 Ubiquitin Protein Ligase 2 using the numbering given in entry NP_008945.2 (GeneID 11060). | |
0.1 mL | |
Cell Biology | |
11060 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction