Learn More
Invitrogen™ WWTR1 Monoclonal Antibody (CL0371)

Description
Immunogen sequence: MNPKPSSWRK KILPESFFKE PDSGSHSRQS STDSSGGHPG PRLAGGAQHV RSHSSPASLQ LGTGAGAAGS PAQQHAHLRQ QSYDVTDELP LPPGWEMTFT ATGQRYFLNH IEKITTWQDP RKAMNQPLNH MNLHPAVSST Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%. Binds to an epitope located within the peptide sequence SSWRKKILPESFFKE as determined by overlapping synthetic peptides.
Specifications
Specifications
| Antigen | WWTR1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | CL0371 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | WWTR1 |
| Gene Accession No. | Q9GZV5 |
| Gene Alias | 2310058J06Rik; 2610021I22Rik; C78399; DKFZp586I1419; FLJ27004; FLJ45718; Tafazzin; TAZ; transcriptional coactivator with PDZ binding motif; transcriptional co-activator with PDZ-binding motif; transcriptional coactivator with PDZ-binding motif; transcriptional co-activator with PDZ-binding motif (TA)Z; transcriptional co-activator with PDZ-binding motif (TAZ); WW domain containing transcription regulator 1; WW domain-containing transcription regulator protein 1; Wwtr1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.