Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WWTR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119927
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119927 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5119927 Supplier Invitrogen™ Supplier No. PA5119927
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: Human placenta tissue lysate, 293T cell lysate, human kidney tissue, mouse brain tissue, A549, HT-29. Subcellular Location: Cell membrane, Cytoplasm, Membrane, Nucleus.

WW domain-containing transcription regulator protein 1 (WWTR1) is a transcriptional coactivator which acts as a downstream regulatory target in the Hippo signaling pathway that plays a pivotal role in organ size control and tumor supression by restricting proliferation and promoting apoptosis. The core of this pathway is composed of a kinase cascade wherein STK3/MST2 and STK4/MST1, in complex with its regulatory protein SAV1, phosphorylates and activates LATS1/2 in complex with its regulatory protein MOB1. In turn, MOB1 phosphorylates and inactivates YAP1 oncoprotein and WWTR1/TAZ. WWTR1 enhances PAX8 and NKX2-1/TTF1-dependent gene activation. WWTR1 regulates the nuclear accumulation of SMADS and has a key role in coupling them to the transcriptional machinery such as the mediator complex. WWTR1 also regulates embryonic stem-cell self-renewal and promotes cell proliferation and epithelial-mesenchymal transition.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WWTR1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene WWTR1
Gene Accession No. Q9EPK5, Q9GZV5
Gene Alias 2310058J06Rik; 2610021I22Rik; C78399; DKFZp586I1419; FLJ27004; FLJ45718; Tafazzin; TAZ; transcriptional coactivator with PDZ binding motif; transcriptional co-activator with PDZ-binding motif; transcriptional coactivator with PDZ-binding motif; transcriptional co-activator with PDZ-binding motif (TA)Z; transcriptional co-activator with PDZ-binding motif (TAZ); WW domain containing transcription regulator 1; WW domain-containing transcription regulator protein 1; Wwtr1
Gene Symbols WWTR1
Host Species Rabbit
Immunogen Recombinant protein within human WWTR1 aa 1-200.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25937, 97064
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.