Learn More
Invitrogen™ WWTR1 Polyclonal Antibody

Description
PA1-4218 detects the TAZ protein in human, mouse, and rat samples. PA1-4218 has been successfully used in Western blot procedures. By Western blot this antibody detects a ~49 kDa protein representing TAZ in MDCK cells. The PA1-4218 immunogen is a synthetic peptide corresponding to residues V(386) E S A L N K S E P FL T W L(400) of the 49kDa human TAZ protein.
Specifications
Specifications
| Antigen | WWTR1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | WWTR1 |
| Gene Accession No. | Q9EPK5, Q9GZV5 |
| Gene Alias | 2310058J06Rik; 2610021I22Rik; C78399; DKFZp586I1419; FLJ27004; FLJ45718; Tafazzin; TAZ; transcriptional coactivator with PDZ binding motif; transcriptional co-activator with PDZ-binding motif; transcriptional coactivator with PDZ-binding motif; transcriptional co-activator with PDZ-binding motif (TA)Z; transcriptional co-activator with PDZ-binding motif (TAZ); WW domain containing transcription regulator 1; WW domain-containing transcription regulator protein 1; Wwtr1 |
| Gene Symbols | WWTR1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.