Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ WZ 4002
GREENER_CHOICESDP

Catalog No. 629050 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
62-905-0 50 mg
62-901-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 62-905-0 Supplier Tocris Bioscience™ Supplier No. 6290/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent mutant-selective EGFR inhibitor

WZ 4002 is a potent irreversible mutant selective EGFR tyrosine kinase inhibitor. It blocks ATP-dependent autophosphorylation of EGFR carrying the T790M or L858R mutation (IC50 = 8 nM). WZ 4002 exhibits selectivity for mutant EGFR over wild-type. In mice bearing T790M mutant lung cancer cell lines WZ 4002 inhibits EGFR, AKT and ERK1/2 phosphorylation and results in significant tumor regression.

Specifications

Chemical Name or Material N-[3-[[5-Chloro-2-[[2-methoxy-4-(4-methyl-1-piperazinyl)phenyl]amino]-4-pyrimidinyl]oxy]phenyl]-2-propenamide
CAS 1213269-23-8
Quantity 50 mg
Target EGF Receptor Inhibitors
Molecular Formula C25H27ClN6O3
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.