Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ X-Gal

Catalog No. p-4530703
Click to view available options
Quantity:
1 g
2 x 1 g

Effectively screen colonies with X-Gal, an inert chromogenic substrate for beta-galactosidase which hydrolyzes X-Gal, forming an intense blue precipitate.

X-Gal (5-bromo-4-chloro-3-indolyl-beta-D-galacto-pyranoside) is an inert chromogenic substrate for beta-galactosidase which hydrolyzes X-Gal into colorless galactose and 4-chloro-3-brom-indigo, forming an intense blue precipitate. Induction of the lacZ gene with IPTG leads to the hydrolysis of X-Gal and to the development of blue colonies.

Induction of the lacZ gene with IPTG leads to the hydrolysis of X-Gal and to the development of blue colonies. X-Gal Solution is a stable, 0.22μm membrane filtered solution formulated for direct use in conjunction with IPTG for blue/white colony screening.
TRUSTED_SUSTAINABILITY

Specifications

Detection Method Colorimetric
Product Type X-Gal
Quantity 1 g
Substrate Properties Chemical Substrate
Substrate Type Beta-Gal Substrate, X-Gal
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.