Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ X-MINI™ Human Selection Kits

Catalog No. p-7192926 Shop All Corning Products
Click to view available options
Label Type:
Biotinylated anti-human CD3 Antibody
Biotinylated anti-human CD8 Antibody

Corning™ X-MINI™ Human Selection Kits are designed for selection of cells and cell subsets from blood and blood products using Buoyancy Activated Cell Sorting (Corning X-BACS™) technology ensuring high purity, consistency and viability of cells for downstream applications.

  • Selected cells are immediately available for downstream applications such as flow cytometry and cell culture expansion.
  • Easy to use
  • Single-use, column-free and gentle on cells
  • High purity: Achieve purities of up to 99%
  • Reproducible results (>86%)
  • High viability (>97%)
  • Process up to 4 x 108 total cells
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) Up to 4 x 108 total cells
For Use With (Application) Blood and Blood Products
Quantity 1/Pk., 1/Cs.
Content And Storage 2°C to 8°C
Includes Biotin Mouse Anti-Human Antibody; Corning X-BACS™ Reagent; Corning X-BACS Buffer; Syringe, 10 mL; 21 G x 1.5 in. Needle; Syringe cap
Label Type Biotinylated anti-human CD8 Antibody
Product Type Cell selection
Detection Method Buoyancy Activated Cell Sorting
Product Line Corning X-BACS
Shipping Condition Room Temperature
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.