Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XAB2 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501726467 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-6467 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-6467 Supplier Proteintech Group Inc Supplier No. 106371AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A novel protein designated XAB2 (XPA-binding protein 2) interacts with XPA, a factor central to both nucleotide excision repair subpathways. Data suggests that XAB2 is a novel component involved in transcription-coupled repair and transcription.

Specifications

Antigen XAB2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.21 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene XAB2
Gene Accession No. Q9HCS7
Gene Alias HCNP, HCRN, KIAA1177, NTC90, Pre mRNA splicing factor SYF1, Protein HCNP, SYF1, XAB2, XPA binding protein 2
Gene Symbols XAB2
Host Species Rabbit
Immunogen XAB2 Fusion Protein Ag0999
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56949
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.