Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ XAF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520090
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520090 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520090 Supplier Invitrogen™ Supplier No. PA520090
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody recognizes multiple isoforms of XAF-1. In Western blot applications, a doublet is detected at ~30kDa. A suggested positive control is human spleen tissue lysate. PA5-20090 can be used with blocking peptide PEP-0208.

The inhibitors of apoptosis (IAP) proteins prevent cell death both in vitro and in vivo by virtue of inhibition of distinct caspases. The X-linked Inhibitor of Apoptosis, XIAP, is a key member of the IAP family of proteins. The caspase inhibiting activity of XIAP is negatively regulated by at least two XIAP-interacting proteins, XIAP-associated factor 1 (XAF1) and Smac/DIABLO. XAF1 binds to XIAP and antagonizes caspase inhibitory activities of XIAP. Expression of XAF1 triggers a redistribution of XIAP from the cytosol to the nucleus. XAF1 is ubiquitously expressed in normal tissues, but is present at low or undetectable levels in many different cancer cell lines.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XAF1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene XAF1
Gene Accession No. Q6GPH4
Gene Alias BIRC4 binding protein; BIRC4-binding protein; Birc4bp; F-box protein 39; Fbox39; HSXIAPAF1; novel TRAF-type zinc finger protein; XAF1; XIAP associated factor 1; XIAP associated factor-1; Xiapaf1; XIAP-associated factor 1
Gene Symbols XAF1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 16 amino acids near the middle of human XAF-1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54739
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.