Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ XAGE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546413
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546413 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546413 Supplier Invitrogen™ Supplier No. PA546413
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: HERHTQTQNH TASPRSPVME SPKKKNQQLK VGILHLGSRQ KKIRIQLRSQ Sequence homology: Human: 100%.

This gene is a member of the XAGE subfamily, which belongs to the GAGE family. The GAGE genes are expressed in a variety of tumors and in some fetal and reproductive tissues. This gene is strongly expressed in Ewing's sarcoma, alveolar rhabdomyosarcoma and normal testis. The protein encoded by this gene contains a nuclear localization signal and shares a sequence similarity with other GAGE/PAGE proteins. Because of the expression pattern and the sequence similarity, this protein also belongs to a family of CT (cancer-testis) antigens. Alternative splicing of this gene, in addition to alternative transcription start sites, results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XAGE1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene XAGE1B
Gene Accession No. Q9HD64
Gene Alias cancer/testis antigen 12.1; cancer/testis antigen family 12, member 1c; cancer/testis antigen family 12, member 1d; cancer/testis antigen family 12, member 1e; cancer/testis associated protein; CT12.1; CT12.1A; CT12.1C; CT12.1D; CT12.1E; CTP9; G antigen family D member 2; GAGED2; protein XAGE-1; X antigen family member 1; X antigen family member 1E; X antigen family, member 1C; X antigen family, member 1D; X antigen family, member 1E; XAGE1; XAGE-1; XAGE1A; XAGE1B; XAGE1C; XAGE1D; XAGE1E
Gene Symbols XAGE1A, XAGE1B
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human XAGE1B.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 653067, 653220
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.