Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ XAGE2 Monoclonal Antibody (4C4)
GREENER_CHOICE

Catalog No. PIMA524437
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524437 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524437 Supplier Invitrogen™ Supplier No. MA524437
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: RNPTPDQKRE DDQGAAEIQV PDLEADLQEL CQTKTGDGCE GGTDVKGKIL PKAEHFKMPE AGEGKSQV.

This gene is a member of the XAGE subfamily, which belongs to the GAGE family. The GAGE genes are expressed in a variety of tumors and in some fetal and reproductive tissues. This gene is strongly expressed in normal testis, and in Ewing's sarcoma, rhabdomyosarcoma, a breast cancer and a germ cell tumor. The protein encoded by this gene shares a sequence similarity with other GAGE/PAGE proteins. Because of the expression pattern and the sequence similarity, this protein also belongs to a family of CT (cancer-testis) antigens.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XAGE2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4C4
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene XAGE2
Gene Accession No. Q96GT9
Gene Alias Cancer/testis antigen 12.2; cancer/testis antigen family 12, member 2; CT12.2; G antigen family D member 3; GAGED3; X antigen family member 2; X antigen family, member 2; X antigen family, member 2B; XAGE2; XAGE-2; XAGE2B
Gene Symbols XAGE2
Host Species Mouse
Immunogen XAGE2 (NP_570133, 44 a.a. ~ 111 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9502
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.