Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
XAGE2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
| Antigen | XAGE2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP16889820
![]() |
Novus Biologicals
NBP16889820UL |
20 μL |
Each for $206.00
|
|
|||||
NBP168898
![]() |
Novus Biologicals
NBP168898 |
100 μL |
Each for $487.50
|
|
|||||
Description
XAGE2 Polyclonal specifically detects XAGE2 in Human samples. It is validated for Western Blot.Specifications
| XAGE2 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Cancer/testis antigen 12.2, cancer/testis antigen family 12, member 2, CT12.2GAGED3, G antigen family D member 3, G antigen, family D, 3, Protein XAGE-2, X antigen family, member 2, XAGE-2, XAGE2B | |
| XAGE2 | |
| IgG | |
| 12 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q96GT9 | |
| 9502 | |
| Synthetic peptides corresponding to XAGE2 (X antigen family, member 2) The peptide sequence was selected from the middle region of XAGE2. Peptide sequence KSRNPTPDQKREDDQGAAEIQVPDLEADLQELCQTKTGDGCEGGTDVKGK. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title