Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
XAGE3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | XAGE3 |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
XAGE3 Polyclonal specifically detects XAGE3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
XAGE3 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
Cancer/testis antigen 12.3, cancer/testis antigen family 12, member 3a, cancer/testis antigen family 12, member 3b, CT12.3, CT12.3a, CT12.3b, G antigen family D member 4, G antigen, family D, 4, MGC71925, PLAC6GAGED4, placenta-specific 6, placenta-specific 6; G antigen, family D, 4, Placenta-specific gene 6 protein, pp9012, Protein XAGE-3, X antigen family, member 3, XAGE-3 | |
XAGE3 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Rabbit | |
Human | |
Q8WTP9 | |
170626 | |
This antibody was developed against a recombinant protein corresponding to amino acids: SQSKTGGECGNGPDDQGKILPKSEQ | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title